7-HC-γ-linolenate ≥98% (by TLC)
Synonyms:
7-Hydroxycoumarinyl-γ-linolenate
BML-ST503-0050
89164-886EA
446.11
USD
89164-886
7-HC-γ-linolenate ≥98% (by TLC)
7-HC-gamma-linolenate
Phospholipase substrate.
PLA₂ fluorogenic substrate for use in assays with phospholipid and Ca²⁺. For use with purified enzyme, as this substrate is nonspecific. Ideal for drug discovery, HTS, and similar applications. Ex.: 360 nm, Em.: 460 nm
Soluble in 100% ethanol (35 mg/ml) or DMSO (50 mg/ml).
Formula:
C₂₇H₃₄O₄ MW: 422.6 g/mol Storage Temperature: Ultra-Low Temperature Freezer (–80°C) |
CAS Number:
161180-12-7 |
Specification Test Results
Purity | ≥98% (TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...