BML-279
Supplier: Enzo Life Sciences
Synonyms: N-[(1-[4-(2-Oxo-3H-benzimidazol-1-yl)-piperidin-1-yl]-ethyl]quinoline-3-carboxamide
BML-ST372-0025 BML-ST372-0005
89164-840EA 431.09 USD
89164-840 89164-838
BML-279
BML-279
PLD inhibitor.
Dual phospholipase D1/D2 (PLD1/PLD2) inhibitor in cellular assays (PLD1 IC50=8 nm in Calu-1 cells; PLD2 IC50=42 nm in HEK293-gfpPLD2 cells).
https://us.vwr.com/stibo/web/std.lang.all/15/41/45401541.jpg