BML-280 ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: N-(2-(4-Oxo-1-phenyl-1,3,8-triazaspiro[4.5]decan-8-yl)-ethyl)quinoline-3-carboxamideVU0285655-1

BML-ST371-0025 BML-ST371-0005
89164-836EA 431.09 USD
89164-836 89164-834
BML-280 ≥98% (by TLC)
BML-280

Potent and selective inhibitor of phospholipase D2 (PLD2) in cellular (PLD1 IC50 = 1900nM Calu-1 cells; PLD2 IC50 = 90nM HEK293-gfpPLD2 cells) and in vitro (hPLD1 IC50 = >20’000nM; hPLD2 IC50 = 500nM) assays.


  • PLD inhibitor


Off-white solid.


Soluble in DMSO (>25mg/ml) or 100% ethanol (>25mg/ml).

Formula: C₂₅H₂₇N₅O₂
MW: 429.5 g/mol
Storage Temperature: Freezer
CAS Number: 1158347-73-9
https://us.vwr.com/stibo/web/std.lang.all/15/40/45401540.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR