Thioetheramide-phosphorylcholine ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: 1-Palmitylthio-2-palmitoylamido-1,2-dideoxy-sn-glycero-3-phosphorylcholine

BML-ST325-0025 BML-ST325-0005
89164-798EA 115.96 USD
89164-798 89164-800
Thioetheramide-phosphorylcholine ≥98% (by TLC)
Thioetheramide-phosphorylcholine

A potent substrate analog inhibitor of snake venom PLA2 (IC50=2 µM). It inhibits human recombinant sPLA2 (IC50=0.9 µM) selectively over cPLA2.


  • Phospholipase inhibitor


White solid.


Soluble in DMSO, 100% ethanol or methanol.

Formula: C₄₀H₈₃N₂O₅PS
MW: 735.6 g/mol
Storage Temperature: Freezer
CAS Number: 116457-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR