Luffariellolide (from Luffariella sp.) ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-ST310-0001
89164-792EA 190.48 USD
89164-792
Luffariellolide (from Luffariella sp.) ≥98% (by TLC)
Luffariellolide

Marine sesterterpenoid. Structurally related to manoalide. Anti-inflammatory phospholipase A2 (PLA2) inhibitor (IC50=0.23µM). Less potent than manoalide, but partially reversible (IC50=0.23µM, bee venom PLA2). Inhibits human recombinant sPLA2 (IC50=9.3µM) selectively over cPLA2.


  • Phospholipase inhibitor


Pale yellow oil.


Soluble in DMSO or 100% ethanol.

Formula: C₂₅H₃₈O₃
MW: 386.6 g/mol
Storage Temperature: Freezer
CAS Number: 111149-87-2
https://us.vwr.com/stibo/web/std.lang.all/15/34/45401534.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR