L-erythro-MAPP ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-SL222-0025
89164-652EA 260.84 USD
89164-652
L-erythro-MAPP ≥98% (by TLC)
L-erythro-MAPP

Negative control for D-erythro-MAPP


Useful as a negative control for D-erythro-MAPP. It does not raise endogenous ceramide levels or inhibit the growth of HL-60 cells.


Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml).


Synonym: (1S,2R-L-erythro-2-N-Myristoylamino)-1-phenyl-1-propanol

Formula: C₂₃H₃₉NO₂
MW: 361.6 g/mol
Storage Temperature: Freezer
CAS Number: 143492-39-1
https://us.vwr.com/stibo/web/std.lang.all/15/26/45401526.jpg

Order Now

Specification Test Results

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR