L-161-982 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-RA118-0005
89162-526EA 404.69 USD
89162-526
L-161-982 ≥98% (by HPLC)
L-161-982

Prostanoid receptor ligand.


A potent and selective prostaglandin EP4 receptor antagonist.


Soluble in DMSO (25 mg/ml).

Formula: C₃₂H₂₉F₃N₄O₄S₂
Storage Temperature: Freezer
CAS Number: 147776-06-5
https://us.vwr.com/stibo/web/std.lang.all/14/90/45401490.jpg

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Identity Determined by ¹H-NMR and MS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR