RWJ-60475 (AM)3
Supplier: Enzo Life Sciences
BML-PR110-0001
89161-778EA 207.18 USD
89161-778
RWJ-60475 (AM)3
RWJ-60475 (AM)3
CD45 inhibitor
RWJ-60475-(AM)3 is a cell-permeable acetoxymethyl (AM) ester of RWJ-60475. The AM esters confer cell membrane permeability and are hydrolyzed by intracellular esterases, trapping the active inhibitor within the cell. The IC50 for the hydrolyzed, active inhibitor is 2 µM.
https://us.vwr.com/stibo/web/std.lang.all/14/75/45401475.jpg