Endothall ≥98%

Supplier: Enzo Life Sciences

BML-PR107-0100
89161-768EA 225.66 USD
89161-768
Endothall ≥98%
Endothall

PP2A inhibitor


Intermediate-potency protein phosphatase 2A (PP2A) inhibitor (IC50=970nM, IC50 for PP1=5000nM). Useful for structure/activity correlations. .

Formula: C₈H₁₀O₅
MW: 186.163 g/mol
MDL Number: MFCD00137362
CAS Number: 145-73-3
https://us.vwr.com/stibo/web/std.lang.all/14/74/45401474.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR