H-Glu-Gly-Arg-CMK
Supplier: Enzo Life Sciences
BML-PI156-0005
89161-704EA 320.75 USD
89161-704
H-Glu-Gly-Arg-CMK
Proteins and Peptides
Protease inhibitor
Potent irreversible inhibitor of uPA (Urokinase), Factor VIIa, Factor IXa, Factor Xa, and trypsin.