L-α-Phosphatidylinositol ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-PH100-0010
89161-548EA
267.12
USD
89161-548
L-α-Phosphatidylinositol ≥98% (by TLC)
L-alpha-Phosphatidylinositol (sodium salt)
Phospholipase substrate.
From soybean. Contains mainly linoleic and palmitic acids. Substrate for phospholipase C.
Soluble in chloroform or ethyl ether.
Formula:
C₄₅H₇₈O₁₃PNa MW: 881.0 g/mol Storage Temperature: Freezer |
CAS Number:
383907-36-6 |
Specification Test Results
Purity | ≥98% (TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...