2,3-Dinor-6-ketoprostaglandin F1α-20,20,20-d3 ≥98% (by TLC), Ultrapure
Supplier: Enzo Life Sciences
|
Danger
|
Synonyms:
2,3-Dinor-6-keto-PGF1α-d3
BML-PG040-0050
89161-512EA
281.64
USD
89161-512
2,3-Dinor-6-ketoprostaglandin F1α-20,20,20-d3 ≥98% (by TLC), Ultrapure
2,3-Dinor-6-ketoprostaglandin F1α-20,20,20-d3
Synonym: 2,3-Dinor-6-keto-PGF₁α-d₃
Formula:
C₁₈H₂₇D₃O₆ MW: 345.5 g/mol Storage Temperature: Freezer |
UN:
1173 ADR: 3,II |
Specification Test Results
Purity | ≥98% (TLC) |
Concentration | 100 µg/ml |
Formulation | Solution in ethyl acetate |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...