Phorbol-12,13-dibutyrate ≥99% (by HPLC)
Supplier: Enzo Life Sciences
|
Warning
|
BML-PE135-0005
89161-386EA
299.57
USD
89161-386
Phorbol-12,13-dibutyrate ≥99% (by HPLC)
Phorbol-12,13-dibutyrate
Widely used PKC activator (10 to 100 ng/ml).
Less potent than phorbol 12-myristate 13-acettate (PMA; TPA), but also less hydrophobic.
Soluble in DMSO (25 mg/ml) or 100% ethanol (20 mg/ml).
Formula:
C₂₈H₄₀O₈ MW: 504.6 g/mol Storage Temperature: Freezer |
CAS Number:
37558-16-0 |
Specification Test Results
Purity | ≥99% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...