Zardaverine ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-PD195-0005 BML-PD195-0025
89161-382EA 518.56 USD
89161-382 89161-380
Zardaverine ≥98% (by HPLC)
Zardaverine

Selective, dual inhibitor of PDE III and PDE IV (IC50s=2.5 and 1.1 µM respectively). Anti-allergic activity, combined inhibition of PDE III and IV have an additive synergistic effect on inhibition of guinea pig bronchospasm.


  • PDE inhibitor


Off-white to yellow solid.


Soluble in DMSO (25mg/ml).

Formula: C₁₂H₁₀F₂N₂O₃
MW: 268.2 g/mol
Storage Temperature: Ambient
CAS Number: 101975-10-4
https://us.vwr.com/stibo/web/std.lang.all/14/16/45401416.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR