Trequinsin hydrochloride ≥98% (by HPLC)
Supplier: Enzo Life Sciences
BML-PD180-0010
89161-370EA
383.07
USD
89161-370
Trequinsin hydrochloride ≥98% (by HPLC)
Trequinsin hydrochloride
Ultrapotent, highly selective inhibitor of cGMP-inhibited phosphodiesterase (PDE lll, IC50=0.3nM) in vitro. Potentiates adenosine-stimulated cAMP accumulation.
- PDE inhibitor
White to pale yellow solid.
Soluble in DMSO (25mg/ml) or 100% ethanol (20mg/ml); slightly soluble in water.
Formula:
C₂₄H₂₇N₃O₃·HCl MW: 405.5 · 36.5 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01076563 CAS Number: 78416-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...