Quazinone ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: Ro 13-6438(R)-6-Chloro-1,5-dihydro-3-methyl- imidazo[2,1-b]quinazolin-2-one

BML-PD170-0010 BML-PD170-0050
89161-356EA 494.86 USD
89161-356 89161-354
Quazinone ≥98% (by HPLC)
Quazinone

Selective inhibitor of cGMP-inhibited phosphodiesterase (PDE III, IC50=0.6µM).


  • PDE inhibitor


White to off-white solid.


Soluble in DMSO (3mg/ml) or 100% ethanol (1mg/ml).

Formula: C₁₁H₁₀ClN₃O
MW: 235.7 g/mol
Storage Temperature: Ambient
CAS Number: 70018-51-8
https://us.vwr.com/stibo/web/std.lang.all/14/10/45401410.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR