RR-src peptide, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-P308-0001
89161-210EA 145.91 USD
89161-210
RR-src peptide, Enzo Life Sciences
Proteins and Peptides
Src kinase substrate
This tyrosine kinase substrate sequence, RRLIEDAEYAARG, was derived from the v-Src autophosphorylation site (original src sequence is IEDNEYTARQG). Km=5 mM in LSTRA cell extracts.