Human Histone H3 (5-23) amide
Supplier: Enzo Life Sciences
BML-P271-0500
89161-188EA 204.06 USD
89161-188
Human Histone H3 (5-23) amide
Proteins and Peptides
HAT substrate
Comprising residues 5-23 of the human histone H3 N-terminal tail, this peptide is centered on Lys14, preferred acetylation site for the GCN5/PCAF family of histone acetyltransferases (HATs). May be used as a substrate for assay of PCAF and hGCN5 HATs.