Axltide,Biotin
Supplier: Enzo Life Sciences
BML-P249-0001
89161-154EA 222.58 USD
89161-154
Axltide,Biotin
Proteins and Peptides
Protein kinase substrate
This peptide, biotin-Ahx-KKSRGDYMTMQIG, is derived from mouse insulin receptor 1 and is a substrate for Axl, DDR2, Mst1, and JAK2 kinases. The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes.