[pSer95, Ala97]-RII (81-99)
Supplier: Enzo Life Sciences
BML-P160-0500
89161-032EA 243.07 USD
89161-032
[pSer95, Ala97]-RII (81-99)
Proteins and Peptides
Calcineurin substrate
This sequence, DLDVPIPGRFDRRVpSVAAE is derived from cAMP-dependent protein kinase regulatory subunit type II [PKA; [Ala-97]-RII (81-99)] with a phosphoserine at Ser95. This peptide can be used directly as a calcineurin (protein phosphatase 2B, PP2B) substrate. Phosphate release can be quantified using the (BML-AK111) detection reagent.