Chk 1 & 2 Substrate
Supplier: Enzo Life Sciences
BML-P158-0001
89161-030EA 279.88 USD
89161-030
Chk 1 & 2 Substrate
Proteins and Peptides
This peptide sequence, KKKVSRSGLYRSPSMPENLNRPR, corresponds to amino acids 206-225 of the phosphatase CDC25C, which is a phosphorylation target of Chk1 on Ser216. The addition of three N-terminal lysines allows the peptide to be used in phosphocellulose kinase assays. Both Chk1 and Chk2 will phosphorylate this peptide.