Pkc Substrate
Supplier: Enzo Life Sciences
BML-P156-0001
89161-026EA 168.58 USD
89161-026
Pkc Substrate
Proteins and Peptides
This peptide sequence, RRFKRQGSFFYFF, was selected from a peptide library for optimal phosphorylation by protein kinase C zeta (PKCζ).