Pkc Substrate
Supplier: Enzo Life Sciences
BML-P155-0500
89161-024EA 187.32 USD
89161-024
Pkc Substrate
Proteins and Peptides
PKC substrate.
Protein kinase C epsilon (PKCε) pseudosubstrate sequence (149-164) but with an Ala to Ser159 substitution (ERMRPRKRQGSVRRRV) which makes it a substrate for phosphorylation. Phospholipid/phorbol ester-dependent substrate for PKC ε, ζ, δ and η.