Protein Tyrosine Phosphatase Substrate
Supplier: Enzo Life Sciences
BML-P153-0001
89161-020EA 159.78 USD
89161-020
Protein Tyrosine Phosphatase Substrate
Proteins and Peptides
This peptide sequence, ENDpYINASL, is derived from a highly conserved region of the T-cell phosphatase, TCPTP, and has been shown to be an excellent general substrate for protein tyrosine phosphatases.