Dibenzepine HCl ≥99%

Supplier: Enzo Life Sciences
Danger

BML-NS107-0100
89160-832EA 84.97 USD
89160-832
Dibenzepine HCl ≥99%
Dibenzepine HCl

Soluble in water.

Formula: C₁₈H₂₁N₃O·HCl
Storage Temperature: Ambient
CAS Number: 315-80-0
https://us.vwr.com/stibo/web/std.lang.all/13/92/45401392.jpg

Order Now

Specification Test Results

Purity ≥99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR