Anpirtoline ≥99% (by HPLC), white solid
Supplier: Enzo Life Sciences
BML-NS100-0025
89160-814EA
684.02
USD
89160-814
Anpirtoline ≥99% (by HPLC), white solid
Anpirtoline
Serotonin receptor ligand.
Serotonin 5-HT₁B receptor agonist.
Soluble in water (25 mg/ml).
Formula:
C₁₀H₁₃ClN₂S·HCl Storage Temperature: Ambient |
CAS Number:
99201-87-3 |
Specification Test Results
Purity | ≥99% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...