HOE-140 ≥95% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
Icatibant
BML-NK104-0001
89160-764EA
357.29
USD
89160-764
HOE-140 ≥95% (by HPLC)
HOE-140
A potent and selective bradykinin B2 antagonist.
HOE-140 is a useful tool for discrimination of the physiological roles of bradykinin B2 versus B1 receptors. Displays a protective effect in myocardial reperfusion injury.
Soluble in water.
Synonym: Icatibant
Formula:
C₅₉H₈₉N₁₉O₁₃S MW: 1304.5 g/mol Storage Temperature: Freezer |
CAS Number:
130308-48-4 |
Specification Test Results
Purity | ≥95% (HPLC) |
Identity | Determined by MS |
Sequence | D-Arg-Arg-Pro-Hyp-Gly-Thi-Ser-Tic-Oic-Arg |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...