R(-)-α-Methylhistamine 2HBr ≥99% (by HPLC)
Supplier: Enzo Life Sciences
BML-NH104-0010
89160-740EA
245.49
USD
89160-740
R(-)-α-Methylhistamine 2HBr ≥99% (by HPLC)
R(-)-α-Methylhistamine 2HBr
Potent and selective histamine H3 agonist. Crosses the blood-brain barrier and inhibits histamine synthesis and release at nanomolar concentrations.
- Histamine receptor ligand
White solid.
Soluble in water (25mg/ml).
Formula:
C₆H₁₁N₃·2HBr MW: 287.0 g/mol Storage Temperature: Ambient |
CAS Number:
868698-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...