A-803467 ≥98%
Supplier: Enzo Life Sciences
Synonyms:
5-(4-Chlorophenyl)-N-(3,5-dimethoxyphenyl)furan-2-carboxamide
BML-NA142-0010
BML-NA142-0050
89160-700EA
115.96
USD
89160-700
89160-702
A-803467 ≥98%
A-803467
Na+ channel blocker
A-803467 is a potent and selective inhibitor of the TTX-resistant voltage-gated sodium channel Nav1.8 which is preferentially expressed on sensory neurons. IC50 for human Nav1.8=8.0 nM (140 nM for rat). It attenuates neuropathic and inflammatory pain in various animal models.
Formula:
C₁₉H₁₆ClNO₄ MW: 357.79 g/mol |
MDL Number:
MFCD10574689 CAS Number: 944261-79-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...