Anti-Neuropeptide Y Rabbit Polyclonal Antibody

NA1233-0100 NA1233-0025
89160-654EA 127.31 USD
89160-654 89160-656
Anti-Neuropeptide Y Rabbit Polyclonal Antibody
Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes human and rat NPY.

Type: Primary
Antigen: NEUROPEPTIDE Y
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR