Benzamil HCl ≥98% (by HPLC), yellow solid
Supplier: Enzo Life Sciences
|
Warning
|
Synonyms:
Benzylamiloride . HCl,
, N-(Benzylamidino)-3,5-diamino-6-chloropyrazinecarboxamide . HCl
BML-NA103-0050
89160-616EA
285.57
USD
89160-616
Benzamil HCl ≥98% (by HPLC), yellow solid
Benzamil HCl
Na+ channel blocker
Derivative of amiloride (Prod. No. BML-CA200). Potent sodium channel blocker.
Soluble in water.
Formula:
C₁₃H₁₄ClN₇O·HCl MW: 356.2 g/mol Storage Temperature: Ambient |
CAS Number:
2898-76-2 |
Specification Test Results
Purity | ≥98% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...