14,15-Leukotriene E4 ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Danger

LPE004-0050
89160-570EA 352.05 USD
89160-570
14,15-Leukotriene E4 ≥98% (by HPLC)
14,15-Leukotriene E4

Novel leukotriene produced via the 15-LO pathway.

Formula: C₂₃H₃₇NO₅S
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
CAS Number: 1000852-57-2
UN: 1992
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/13/63/45401363.jpg

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Concentration 50 µg/ml
Formulation Oil dissolved in methanol/ammonium acetate buffer, 70/30, pH 5.6

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR