Phosphatidic acid, dipalmitoyl ≥98% (by TLC), white solid

Supplier: Enzo Life Sciences

Synonyms: 1,2-Dipalmitoyl-sn-glycero-3-phosphate . sodium saltDPPA

BML-LP103-0500 BML-LP103-0100
89160-544EA 146.98 USD
89160-544 89160-546
Phosphatidic acid, dipalmitoyl ≥98% (by TLC), white solid
Phosphatidic acid, dipalmitoyl

PKC activator


A mitogenic lipid phosphate produced via the action of phospholipase D. Mediates agonist-dependent Raf-1 translocation and activation of the MAP kinase cascade.


Fine suspension in ethanol (5 mg/ml) with 15 minutes of sonication at room temperature.

Formula: C₃₅H₆₈O₈P·Na
Storage Temperature: Freezer
CAS Number: 169051-60-9
https://us.vwr.com/stibo/web/std.lang.all/13/48/45401348.jpg

Order Now

Specification Test Results

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR