N-Acetyl-leukotriene E4 ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Danger

LEA004-0050
89160-528EA 359.07 USD
89160-528
N-Acetyl-leukotriene E4 ≥98% (by HPLC)
N-Acetyl-leukotriene E4

Major bilary metabolite of cysteinyl leukotrienes in the rat. Approximately 100-fold less active than LTC4. Equiactive with LTE4 in guinea pig lung parenchyma assay.


Soluble in ethanol.

Formula: C₂₅H₃₉NO₆S
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
CAS Number: 80115-95-3
UN: 1992
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/13/44/45401344.jpg

Order Now

Specification Test Results

Concentration 50 µg/ml
Purity ≥98% (HPLC)
Formulation Solution in methanol/ammonium acetate buffer, 70:30.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR