1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine ≥98% (by TLC)
Supplier: Enzo Life Sciences
Synonyms:
Carbamyl-PAF
BML-L120-0005
89160-446EA
406.31
USD
89160-446
1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine ≥98% (by TLC)
1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine
PAF receptor activator.
Biologically potent, non-metabolisable analog of platelet-activating factor (PAF). Induces half-maximal PMNL degranulation response at 16 nM and binds stereospecifically to high and low affinity PAF receptors. Completely resists inactivation by acetylhydrolase.
Soluble in 100% ethanol.
Formula:
C₂₆H₅₅N₂O₇P MW: 538.707 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00065891 CAS Number: 91575-58-5 |
Specification Test Results
Purity | ≥98% (TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...