ET-18-OCH3 ≥98% (by TLC)
Supplier: Enzo Life Sciences
Synonyms:
1-O-Octadecyl-2-O-methyl-sn-glycero-3-phosphocholine
Produces strong antitumor and antimetastatic activities. Inhibits phosphatidylinositol specific phospholipase C (PLC, IC50=0.4-9.6μM) and protein kinase C from various leukemic cells. Inhibits CoA-independent transacylase (IC50=0.5μM). Induces apoptosis in HeLa cells and HL-60 cells. Antineoplastic activity.
- Antitumor agent
White waxy solid.
Soluble in 100% ethanol (15mg/ml), DMSO or dimethyl formamide.
Formula:
C₂₇H₅₈NO₆P MW: 523.736 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00063527 CAS Number: 77286-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...