Nicotinamide

Supplier: Enzo Life Sciences

Synonyms: Niacinamidepyridine-3-carboxamideVitamin B33-(Aminocarbonyl)pyridine

BML-KI283-0500
89160-364EA 50.01 USD
89160-364
Nicotinamide
Nicotinamide

Nicotinamide, the water-soluble amide of nicotinic acid, is a component of the two most important coenzymes-nicotinamide adenine dinucleotide and nicotinamide adenine dinucleotide phosphate. Nicotinamide is involved in numerous oxidation-reduction reactions in mammalian biological systems acting as an antioxidant.


Soluble in water, 100% ethanol or glycerol. Slightly soluble in chloroform, almost insoluble in ether or acetone.

Formula: C₆H₆N₂O
MW: 122.13 g/mol
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
MDL Number: MFCD00006395
CAS Number: 98-92-0
EINECS: 202-713-4
https://us.vwr.com/stibo/web/std.lang.all/13/28/45401328.jpg

Order Now

Specification Test Results

Formulation 50 mM nicotinamide in 50 mM tris/Cl, pH 8.0, 137 mM NaCl, 2.7 mM KCl, 1 mM MgCl₂.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR