Dendrotoxin-K (Dendroaspis P. Polylepis)
Supplier: Enzo Life Sciences
BML-KC164-0070
89160-254EA 678.79 USD
89160-254
Dendrotoxin-K (Dendroaspis P. Polylepis)
Proteins and Peptides
Ultra-pure
Blocks mainly Kv1.1 (EC50=0.6nM) but also Kv1.2 channels.