15-Ketoeicosa-6E,8Z,11Z,13E-tetraenoic acid ≥95% (by HPLC)
Supplier: Enzo Life Sciences
|
Danger
|
BML-K015-0050
89160-194EA
225.66
USD
89160-194
15-Ketoeicosa-6E,8Z,11Z,13E-tetraenoic acid ≥95% (by HPLC)
15-Ketoeicosa-6E,8Z,11Z,13E-tetraenoic acid
Biosynthesis via 15-OH prostaglandin dehydrogenase from lung and liver microsomes and by a nonenzymatic process.
Storage Temperature:
Ultra-Low Temperature Freezer (–80°C) |
CAS Number:
81416-72-0 UN: 1170 ADR: 3,II |
Specification Test Results
Purity | ≥95% (HPLC) |
Concentration (for 50 µg size) | 50 µg/ml for 50 µg size |
Concentration (for 1 mg size) | 1 mg/mL |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...