5-Ketoeicosa-6E,8Z,11Z,14E-tetraenoic acid ≥95% (by HPLC)
Supplier: Enzo Life Sciences
BML-K005-0050
89160-190EA
240.27
USD
89160-190
5-Ketoeicosa-6E,8Z,11Z,14E-tetraenoic acid ≥95% (by HPLC)
5-Ketoeicosa-6E,8Z,11Z,14E-tetraenoic acid
R527 ligand.
Stimulates neutrophil degranulation and Ca²⁺ mobilisation by a mechanism independent of the LTB4 receptor. Has been shown to act as an agonist at a G protein-coupled receptor (R527). Induces the infiltration of granulocytes into human skin.
Formula:
C₂₀H₃₀O₃ Storage Temperature: Ultra-Low Temperature Freezer (–80°C) |
CAS Number:
106154-18-1 UN: 1170 ADR: 3,II |
Specification Test Results
Concentration | 50 µg/ml |
Formulation | Oil dissolved in ethanol |
Purity | ≥95% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...