α-Hydroxytriazolam ≥96% (by TLC)
Supplier: Enzo Life Sciences
Warning
|
BML-J130-0001
89160-180EA
182.14
USD
89160-180
α-Hydroxytriazolam ≥96% (by TLC)
alpha-Hydroxytriazolam
An active but short-lived metabolite of triazolam.
Soluble in DMSO (5 mg/mL warm) or methanol (1 mg/mL warm).
Formula:
C₁₇H₁₂Cl₂N₄O MW: 359.2 g/mol Storage Temperature: Freezer |
CAS Number:
37115-45-0 |
Specification Test Results
Purity | ≥96% (TLC) |
Identity | Determined by H-NMR and MS |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...