α-Hydroxyalprazolam ≥98% (by TLC)
Supplier: Enzo Life Sciences
Warning
|
BML-J125-0005
89160-176EA
460.98
USD
89160-176
α-Hydroxyalprazolam ≥98% (by TLC)
alpha-Hydroxyalprazolam
Metabolite of alprazolam.
Soluble in DMSO (2.5 mg/mL) or ethanol (2.5 mg/mL).
Caution: Store, as supplied, at room temperature for up to 1 year. Store solutions at -20 °C for up to 3 months.
Formula:
C₁₇H₁₃CIN₄O MW: 324.8 g/mol Storage Temperature: Ambient |
CAS Number:
37115-43-8 |
Specification Test Results
Purity | ≥98% (TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...