Anti-ITGB1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
IG6061-0100
89160-142EA 265.01 USD
89160-142
Anti-ITGB1 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Western Blot

Recognizes human beta1 integrin.

Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR