5(S)-Hydroperoxyeicosa-6E,8Z,11Z,14Z-tetraenoic acid (HPETE)

Supplier: Enzo Life Sciences
Danger

BML-HP005-0050
89160-092EA 244.45 USD
89160-092
5(S)-Hydroperoxyeicosa-6E,8Z,11Z,14Z-tetraenoic acid (HPETE)
5(S)-Hydroperoxyeicosa-6E,8Z,11Z,14Z-tetraenoic acid (HPETE)

Augments histamine release and calcium flux.


Primary metabolite of arachidonic acid via the 5-lipoxygenase pathway. Intermediate in leukotriene biosynthesis. Augments histamine release and calcium flux.

Formula: C₂₀H₃₂O₄
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
CAS Number: 71774-08-8
UN: 1170
ADR: ,II

Order Now

Specification Test Results

Source Synthetic
Formulation 50 µg/mL in ethanol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR