20-Hydroxyleukotriene B4 ≥99% (by HPLC)
Supplier: Enzo Life Sciences
Danger
|
HLB004-0050
89160-072EA
497.69
USD
89160-072
20-Hydroxyleukotriene B4 ≥99% (by HPLC)
20-Hydroxyleukotriene B4
Primary LTB4 metabolite produced by ω oxidation in PMNL retaining some chemotactic activity. ω oxidation is blocked by 17-ODYA.
Formula:
C₂₀H₃₂O₅ Storage Temperature: Ultra-Low Temperature Freezer (–80°C) |
CAS Number:
79516-82-8 UN: 1170 ADR: ,II |
Specification Test Results
Source | Synthetic |
Concentration | 50 µg/mL |
Formulation | Solution in ethanol |
Purity | ≥99% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...