12(R)-Hydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (HETE) ≥99%
Supplier: Enzo Life Sciences
Synonyms:
12(R)-HETE
Biosynthesis in psoriatic skin and corneal epithelial cells via cytochrome P-450. More potent than 12(S)-HETE in inducing human leukocyte chemokinesis and aggregation. Proinflammatory effects in human skin. Inhibits renal, cardiac and corneal Na+,K+-ATPase. Reduces intraocular pressure.
- ATPase inhibitor
Biosynthesis in psoriatic skin and corneal epithelial cells via cytochrome P-450. More potent than 12(S)-HETE in inducing human leukocyte chemokinesis and aggregation. Proinflammatory effects in human skin. Inhibits renal, cardiac and corneal Na+,K+-ATPase. Reduces intraocular pressure.
Formula:
C₂₀H₃₂O₃ MW: 320.5 g/mol Storage Temperature: Ultra-Low Temperature Freezer (–80°C) |
CAS Number:
82337-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...