15(S)-Hydroxyeicosa-5Z,8Z,11Z,13E-tetraenoic acid (HETE) ≥99% (by HPLC)
Supplier: Enzo Life Sciences
Danger
|
BML-H015-1000
BML-H015-0050
89160-024EA
103.77
USD
89160-024
89160-026
15(S)-Hydroxyeicosa-5Z,8Z,11Z,13E-tetraenoic acid (HETE) ≥99% (by HPLC)
15(S)-Hydroxyeicosa-5Z,8Z,11Z,13E-tetraenoic acid (HETE)
Biosynthesis in human neutrophils, eosinophils and reticulocytes. Most abundant eicosanoid in human bronchi. Regulates T lymphocyte mitogenesis. Induces immunosupression. Stimulates migration of capillary endothelial cells and neovascularisation.
Storage Temperature:
Ultra-Low Temperature Freezer (–80°C) |
CAS Number:
54845-95-3 UN: 1170 ADR: ,II |
Specification Test Results
Source | Synthetic |
Formulation | Oil dissolved in ethanol |
Purity | ≥99% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...