8(S)-Hydroxyeicosa-5Z,9E,11Z,14Z-tetraenoic acid (HETE) solution 50 µg/ml in ethanol , 99% (by HPLC)
Supplier: Enzo Life Sciences
|
Danger
|
BML-H008-0050
89160-016EA
236.16
USD
89160-016
8(S)-Hydroxyeicosa-5Z,9E,11Z,14Z-tetraenoic acid (HETE) solution 50 µg/ml in ethanol , 99% (by HPLC)
8(S)-Hydroxyeicosa-5Z,9E,11Z,14Z-tetraenoic acid (HETE)
Activator of PKC and PPARα.
Biosynthesis in human neutrophils, phorbol ester-treated mouse epidermis and by hepatic microsomal cytochrome P-450. Chemotactic for neutrophils. Activator of PPARα. Activates mouse skin protein kinase C (PKC).
Formula:
C₂₀H₃₂O₃ Storage Temperature: Ultra-Low Temperature Freezer (–80°C) |
CAS Number:
98462-03-4 UN: 1170 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...