Anti-GNRH Guinea Pig Polyclonal Antibody

Supplier: Enzo Life Sciences
GZ1110-0025
89160-012EA 280.56 USD
89160-012
Anti-GNRH Guinea Pig Polyclonal Antibody
Antibodies
Host: Guinea pig

  • For Immunohistochemistry

Recognizes human, mouse, rat, and cow GnRH.

Type: Primary
Antigen: GONADOTROPIN RELEASING HORMONE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Guinea Pig
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR