Anti-GNRH Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
GZ1092-0025
89160-010EA 498.9 USD
89160-010
Anti-GNRH Mouse Monoclonal Antibody
Antibodies
Host: Mouse

  • For Immunohistochemistry

Recognizes human, rat, cow and guinea pig GnRH.

Type: Primary
Antigen: GONADOTROPIN RELEASING HORMONE
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR